EMAIL THIS PAGE TO A FRIEND
European Journal of Pharmacology
Is there a trigger role of peroxynitrite in the anti-arrhythmic effect of ischaemic preconditioning and peroxynitrite infusion
PMID 21699894