EMAIL THIS PAGE TO A FRIEND
Journal of Controlled Release : Official Journal of the Controlled Release Society
Viral genome dNAlipoplexes elicit in situ oncolytic viral replication and potent antitumor efficacy via systemic delivery.
PMID 21699929