EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Study on the fluorescence enhancement in Lanthanum(III)-carminic acid-cetyltrimethylammonium bromide system and its analytical application.
PMID 21703912
Abstract
A fluorescent enhancement system carminic acid (CA)-La3+-CTAB is found and based on this finding a new fluorimetric method for the determination of CA is developed. Under optimized conditions, the enhanced intensities of fluorescence are quantitatively in proportion to the concentrations of CA in the range of 0.01231-12.31 μg mL(-1). The detection limit is 10.92 ng mL(-1). Compared with other methods that have been reported to determine CA, this method has high sensitivity, stability and wide linear range. In addition, the luminescence mechanism indicates that the complex of La3+-CA (1:2) forms and solubilizes in CTAB micelle.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
229253
Carminic acid
C22H20O13
11298
Carminic acid, analytical standard
C22H20O13