EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Sulfo-N-hydroxysuccinimide interferes with bicinchoninic acid protein assay.
PMID 21704014
Abstract
This study revealed a major interference from sulfo-N-hydroxysuccinimide (sulfo-NHS) in the bicinchoninic acid (BCA) protein assay. Sulfo-NHS, a common reagent used in bioconjugation and analytical biochemistry, exhibited absorbance signals and absorbance peaks at 562 nm, comparable to bovine serum albumin (BSA). However, the combined absorbance of sulfo-NHS and BSA was not strictly additive. The sulfo-NHS interference was suggested to be caused by the reduction of Cu(2+) in the BCA Kit's reagent B (4% cupric sulfate) in a manner similar to that of the protein.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14335
2,2′-Biquinoline-4,4′-dicarboxylic acid, ≥90% (TLC)
C20H12N2O4