EMAIL THIS PAGE TO A FRIEND
British journal of clinical pharmacology
Short QT syndrome in infancy. Therapeutic drug monitoring of hydroquinidine in a newborn infant.
PMID 21707701
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
254819
Hydroquinidine hydrochloride, 98%
C20H26N2O2 · HCl
337714
Hydroquinine, 98%
C20H26N2O2