EMAIL THIS PAGE TO A FRIEND
Journal of clinical oncology : official journal of the American Society of Clinical Oncology
Clinical development of vascular disrupting agents: what lessons can we learn from ASA404?
PMID 21709201