EMAIL THIS PAGE TO A FRIEND
Health physics
An emergency bioassay method for actinides in urine.
PMID 21709501
Abstract
A rapid bioassay method has been developed for the sequential measurements of actinides in human urine samples. The method involves actinide separation from a urine matrix by co-precipitation with hydrous titanium oxide (HTiO), followed by anion exchange and extraction chromatography column purification, and final counting by alpha spectrometry after cerium fluoride micro-precipitation. The minimal detectable activities for the method were determined to be 20 mBq L(-1) or less for plutonium, uranium, americium and curium isotopes, with an 8-h sample turn-around time. Spike tests showed that this method would meet the requirements for actinide bioassay following a radiation emergency.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
229555
Cerium(III) fluoride, anhydrous, powder, 99.99% trace metals basis
CeF3