EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Chemical evolution of a bacterium's genome.
PMID 21710668
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
224588
5-Chlorouracil, ReagentPlus®, 99%
C4H3ClN2O2