EMAIL THIS PAGE TO A FRIEND
Herpesviridae
Detection of human cytomegalovirus in motile spermatozoa and spermatogenic cells in testis organotypic culture.
PMID 21711549
Abstract
The presence of human cytomegalovirus (HCMV) in male genital tract suggests its vertical transmission with spermatozoa and the development of a potentially dangerous fetal infection. The objective of the present study was to evaluate the possibility of intracellular HCMV localization in male germ cells and to examine the effect of the virus on human spermatogenesis. Semen samples from 91 infertile and 47 fertile men were analyzed. HCMV was detected by real time PCR, rapid culture method and PCR in situ. Human testis organotypic culture and quantitative karyological analysis were used to investigate viral effects on spermatogenesis. Localization of HCMV in immature germ cells and spermatozoa was studied by immunostaining with monoclonal antibodies and ultrastructural analysis of infected organotypic culture. Viral DNA was detected in 12.3% samples of motile spermatozoa, while infectious activity only in 2.9% infertile and fertile men without statistically significant intergroup difference. According to PCR in situ, the mean percentage of infected cell in both groups was 1.5% (0.25%-15%), which can serve as a criterion for evaluating the risk of HCMV transmission. In HCMV-infected organotypic culture viral antigens were identified in spermatides on day 4, in spermatogonia and spermatocytes on day 8, and in spermatozoa on day 14. Empty and full capsides and virions were visualized in germ cells by electron microscopy. The number of cells before introduction in culture was taken for 100%. On day 14 infected culture contained 36.8% spermatogonia, 18.7% spermatocytes, 27.6% round spermatides and 42.5% elongated spermatides; in comparison with 82.2%, 51.5%, 70.4% and 65.7% in uninfected culture, respectively (all p < 0.05). There were no changes in the number and viability of spermatozoa. HCMV was detected in male germ cells, both in sperm samples and in testis organotypic culture. The virus may infect immature germ cells which develop to mature HCMV-carrying spermatozoa. A considerable decrease in the number of immature germ cells indicates that HCMV produces a direct gametotoxic effect and can contribute to male infertility.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05470
Bovine serum albumin, lyophilized powder, crystallized, ≥98.0% (GE)
A7888
Bovine Serum Albumin, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
A9647
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
A3294
Bovine Serum Albumin, heat shock fraction, protease free, pH 7, ≥98%
A3059
Bovine Serum Albumin, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
A0281
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
A7638
Bovine Serum Albumin, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
A8022
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
A3912
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
A3803
Bovine Serum Albumin, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
B4287
Bovine Serum Albumin, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
A8806
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
A4378
Bovine Serum Albumin, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)
A7511
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥97% (agarose gel electrophoresis), 1X Crystallized
A4919
Bovine Serum Albumin, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%
A3311
Bovine Serum Albumin, powder, BioXtra
A1933
Bovine Serum Albumin, chromatographically purified, New Zealand origin, low endotoxin, suitable for cell culture, pH 7, ≥98%
A4161
Bovine Serum Albumin, lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture
A9430
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
A2934
Bovine Serum Albumin, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)
B6917
Bovine Serum Albumin, lyophilized powder, suitable for (for molecular biology), Non-acetylated
A3156
Bovine Serum Albumin, lyophilized powder, γ-irradiated, Globulin Free, BioXtra, suitable for cell culture
A9306
Bovine Serum Albumin, lyophilized powder, low endotoxin, ≥95% (agarose gel electrophoresis)
A9543
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
A3675
Bovine Serum Albumin, lyophilized powder, low endotoxin, ≥98% (agarose gel electrophoresis)
A9056
Bovine Serum Albumin, lyophilized powder, Microbiologically tested, ≥96% (agarose gel electrophoresis)
A9085
Bovine Serum Albumin, lyophilized powder, essentially IgG free, ≥96% (agarose gel electrophoresis)
A7517
Bovine Serum Albumin, For use as a marker in SDS-PAGE
A1470
Bovine Serum Albumin, lyophilized powder, low endotoxin, Cohn Analog, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
A8654
Bovine Serum Albumin, dimer mol wt ~132 kDa