EMAIL THIS PAGE TO A FRIEND
The Journal of antimicrobial chemotherapy
In vitro susceptibility of Actinobaculum schaalii to mecillinam.
PMID 21712238
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33447
Mecillinam, VETRANAL®, analytical standard
C15H23N3O3S