EMAIL THIS PAGE TO A FRIEND
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology
Reduction of isoviolanthrone by reaction with photogenerated radicals.
PMID 21713275
Abstract
The properties of isoviolanthrone (V(1)) and its corresponding dihydro form (V(1)-H(2)) in argon-saturated 1-phenylethanol in the presence of acetophenone were examined by photophysical and photochemical methods. Upon photolysis of the sensitizer, a radical-induced reduction of V(1) to V(1)-H(2) occurs. The quantum yield of this reduction is close to unity. The fluorescence of V(1) has a quantum yield of 0.08, for V(1)-H(2)Φ(f) is strongly enhanced and the emission maximum is blue-shifted. The ketyl radical, the semiquinone radical and V(1)-H(2) were observed in this sequence by flash photolysis. V(1)-H(2) is unstable and undergoes oxidation to V(1) on admission of oxygen.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P13800
1-Phenylethanol, 98%
C8H10O
W268518
α-Methylbenzyl alcohol, ≥99%, FCC, FG
C8H10O