EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
Antitrypanosomal therapy for chronic Chagas' disease.
PMID 21714649
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N3415
Nifurtimox, ≥98% (HPLC)
C10H13N3O5S