EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Plasma esterases in the tegu lizard Tupinambis merianae (Reptilia, Teiidae): impact of developmental stage, sex, and organophosphorus in vitro exposure.
PMID 21717171
Abstract
In this study, we determined normal serum butyrylcholinesterase (BChE) and carboxylesterase (CbE) activities in Tupinambis merianae in order to obtain reference values for organophosphorus pesticide monitoring. Forty-two T. merianae individuals were grouped by sex and size to identify potential differences in their enzyme levels to allow for proper representation of normal values for females, males, juveniles, and hatchlings. Mean CbE was determined using two model substrates: alpha-naphtylacetate (α-NA) and p-nitrophenyl valerate (4-NPV). BChE and CbE sensitivity to malaoxon (Mx) was also evaluated as well as the possibility of BChE reactivation with pyridine-2-aldoxime methochloride (2-PAM). Mean adult females' BChE was significantly higher than adult males, juveniles, and hatchlings. No significant differences were found between groups regarding CbE. CbE (4-NPV) activity showed slightly negative correlation with lizard snout-vent length, while BChE and CbE (α-NA) showed no correlation with body size. Apparent IC(50) values for BChE and CbE (α-NA) suggested different sensitivities among groups. CbE (4-NPV) could not be inhibited. All Mx-inhibited groups treated with 2-PAM in a final concentration of 2.8 mM showed clear signs of reactivation. In conclusion, the results demonstrate that (1) plasma esterase activity did not vary with age and sex, except for BChE activity, and (2) because biological and environmental variables could be confounding factors in the response of plasma cholinesterases, complementary biomarkers like CbE inhibition and oxime-induced reactivation of esterases are strongly recommended.
Related Materials