EMAIL THIS PAGE TO A FRIEND
Annals of nuclear medicine
Internal radiotherapy techniques using radiolanthanide praseodymium-142: a review of production routes, brachytherapy, unsealed source therapy.
PMID 21720780
Abstract
Radionuclides of rare earth elements are gaining importance as emerging therapeutic agents in nuclear medicine. β(-)-particle emitter 142Pr [T (1/2) = 19.12 h, E(-)β = 2.162 MeV (96.3%), Eγ = 1575 keV (3.7%)] is one of the praseodymium-141 (100% abundant) radioisotopes. Production routes and therapy aspects of 142Pr will be reviewed in this paper. However, 142Pr produces via 141Pr(n, γ) 142Pr reaction by irradiation in a low-fluence reactor; 142Pr cyclotron produced, could be achievable. 142Pr due to its high β(-)-emission and low specific gamma γ-emission could not only be a therapeutic radionuclide, but also a suitable radionuclide in order for biodistribution studies. Internal radiotherapy using 142Pr can be classified into two sub-categories: (1) unsealed source therapy (UST), (2) brachytherapy. UST via 142Pr-HA and 142Pr-DTPA in order for radiosynovectomy have been proposed. In addition, 142Pr Glass seeds and 142Pr microspheres have been utilized for interstitial brachytherapy of prostate cancer and intraarterial brachytherapy of arteriovenous malformation, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
263176
Praseodymium, powder, ~40 mesh, 99.5% trace rare earth metals basis
Pr
261173
Praseodymium, ingot, under oil, 99.9% trace rare earth metals basis
Pr
GF91191191
Praseodymium, foil, 25x25mm, thickness 0.25mm, as rolled, 99%
Pr
GF98899687
Praseodymium, foil, 50x50mm, thickness 0.25mm, as rolled, 99%
Pr
GF96026531
Praseodymium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Pr
GF75299170
Praseodymium, foil, not light tested, 25x25mm, thickness 0.015mm, as rolled, 99%
Pr
GF69747621
Praseodymium, foil, not light tested, 25x25mm, thickness 0.01mm, as rolled, 99%
Pr
GF82608295
Praseodymium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Pr
GF34755857
Praseodymium, foil, not light tested, 50x50mm, thickness 0.005mm, as rolled, 99%
Pr
GF92630246
Praseodymium, foil, not light tested, 50x50mm, thickness 0.015mm, as rolled, 99%
Pr
GF48004804
Praseodymium, foil, not light tested, 50x50mm, thickness 0.01mm, as rolled, 99%
Pr
GF66110329
Praseodymium, rod, 100mm, diameter 6.35mm, cast, 99%
Pr
GF06128883
Praseodymium, rod, 50mm, diameter 6.35mm, cast, 99%
Pr
GF50132191
Praseodymium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Pr
GF66524643
Praseodymium, wire reel, 200mm, diameter 1.0mm, hard, 99.9%
Pr
GF07601059
Praseodymium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Pr