EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Highly selective fluorescent probe for vicinal-dithiol-containing proteins and in situ imaging in living cells.
PMID 21721084
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3075
Phenylarsine oxide, ≥97%, powder
C6H5AsO
402494
Phenylarsine oxide, technical grade
C6H5AsO