EMAIL THIS PAGE TO A FRIEND
Natural product research
Alkaloids and flavonoids of Argemone mexicana.
PMID 21722040
Abstract
A new protopine alkaloid, protomexicine and a new isoflavonoid, mexitin, together with 8-methoxydihydrosanguinarine, 13-oxoprotopine, rutin and quercetrin have been isolated from the aerial part of the methanolic extract of Argemone mexicana. The structures of these compounds have been established by various spectral data. The structure of protomexicine was further confirmed by heteronuclear multiple bond correlation experiment. Protomexicine and mexitin are new compounds, and other compounds are first reported from A. mexicana and genus Argemone.