EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Fused indolines by palladium-catalyzed asymmetric C-C coupling involving an unactivated methylene group.
PMID 21726030
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
743259
1,3-Bis[(1S)-2,2-dimethyl-1-(1-naphthyl)propyl]imidazolinium iodide, ≥96% (HPLC)
C33H39IN2