EMAIL THIS PAGE TO A FRIEND
ACS applied materials & interfaces
Adhesion promotion via noncovalent interactions in self-healing polymers.
PMID 21728372
Abstract
Dimethylnorbornene ester (DNE) is successfully used as a noncovalent adhesion promoter. DNE was confirmed to copolymerize with dicyclopentadiene (DCPD) to yield a copolymer with better adhesion to an EPON 828 epoxy matrix relative to poly(DCPD) alone. The mechanical properties of the copolymer were comparable to that of poly(DCPD) alone. An optimized blend of the monomers was encapsulated using a urea-formaldheyde microencapsulation procedure and the resulting capsules were used for in situ self-healing experiments. Improved healing efficiency was observed for samples containing the DCPD/DNE capsules under conditions in which the monomers were efficiently polymerized.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
454338
Dicyclopentadiene, contains BHT as stabilizer
C10H12