EMAIL THIS PAGE TO A FRIEND
Epilepsia
Effect of intracranial administration of ethosuximide in rats with spontaneous or pentylenetetrazol-induced spike-wave discharges.
PMID 21729038
Abstract
Generalized absence seizures are characterized by bilateral spike-wave discharges (SWDs), particularly in the frontoparietal cortical region. In WAG/Rij and GAERS rats with absence epilepsy, recent evidence indicates that SWDs arise first from the lateral somatosensory cortex (LSC), that is, the cortical focus theory. To further understand the cortical role in SWD generation, two epileptic rat models were assessed. Two models, Long-Evans rats with spontaneous SWDs and Wistar rats with low-dose pentylenetetrazol-induced SWDs (20 mg/kg, i.p.), were administered intracortical or intrathalamic ethosuximide (ESM) or saline. Electroencephalographic recordings were analyzed before and after intracranial microinfusion to evaluate onset, frequency, and duration of SWDs. In both epileptic rat models, ESM in the LSC significantly reduced SWD number, shortened SWD duration, and delayed SWD onset compared to saline. By contrast, ESM in the medial somatosensory cortex had little effect compared to saline. Intrathalamic infusion of ESM only delayed SWD onset. These findings suggest that the LSC may be essential for the occurrence of SWDs. Our data support the cortical focus theory for the generation of absence seizures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E7138
Ethosuximide
C7H11NO2