EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
New regimens to prevent tuberculosis in adults with HIV infection.
PMID 21732833
Abstract
Treatment of latent tuberculosis in patients infected with the human immunodeficiency virus (HIV) is efficacious, but few patients around the world receive such treatment. We evaluated three new regimens for latent tuberculosis that may be more potent and durable than standard isoniazid treatment. We randomly assigned South African adults with HIV infection and a positive tuberculin skin test who were not taking antiretroviral therapy to receive rifapentine (900 mg) plus isoniazid (900 mg) weekly for 12 weeks, rifampin (600 mg) plus isoniazid (900 mg) twice weekly for 12 weeks, isoniazid (300 mg) daily for up to 6 years (continuous isoniazid), or isoniazid (300 mg) daily for 6 months (control group). The primary end point was tuberculosis-free survival. The 1148 patients had a median age of 30 years and a median CD4 cell count of 484 per cubic millimeter. Incidence rates of active tuberculosis or death were 3.1 per 100 person-years in the rifapentine-isoniazid group, 2.9 per 100 person-years in the rifampin-isoniazid group, and 2.7 per 100 person-years in the continuous-isoniazid group, as compared with 3.6 per 100 person-years in the control group (P>0.05 for all comparisons). Serious adverse reactions were more common in the continuous-isoniazid group (18.4 per 100 person-years) than in the other treatment groups (8.7 to 15.4 per 100 person-years). Two of 58 isolates of Mycobacterium tuberculosis (3.4%) were found to have multidrug resistance. On the basis of the expected rates of tuberculosis in this population of HIV-infected adults, all secondary prophylactic regimens were effective. Neither a 3-month course of intermittent rifapentine or rifampin with isoniazid nor continuous isoniazid was superior to 6 months of isoniazid. (Funded by the National Institute of Allergy and Infectious Diseases and others; ClinicalTrials.gov number, NCT00057122.).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R0533
Rifapentine
C47H64N4O12