Journal of veterinary pharmacology and therapeutics
Preliminary pharmacokinetics and cardiovascular effects of fenoldopam continuous rate infusion in six healthy dogs.
PMID 21732952
Fenoldopam is a selective dopamine-1 receptor agonist that causes peripheral arterial vasodilation, increased renal blood flow, and diuresis. Enthusiasm exists for the use of fenoldopam in nonpolyuric kidney injury in dogs, although pharmacokinetic data are lacking. The purpose of this study was to collect basic pharmacokinetic and hemodynamic effect data for fenoldopam when administered to healthy awake dogs. Six healthy, awake beagles were given a 180-min fenoldopam constant rate infusion at 0.8 μg/kg per minute followed by a 120-min washout period. Citrated blood was collected during and after infusion for the measurement of plasma fenoldopam concentration by HPLC with mass spectrometry. Heart rate and indirect systolic blood pressure were concurrently measured. Mean ± SD, steady-state plasma fenoldopam concentrations of 20 ± 17 ng/mL were achieved within 10 min of starting the infusion. Area under the plasma concentration-time curve was 3678 ± 3030 ng/mL · min, and plasma clearance was 66 ± 43 mL/min per kg. Elimination was rapidly achieved in all dogs. Heart rate and systolic blood pressure were unaffected by the fenoldopam infusion. Based on the results of this study, further evaluation of the effects of fenoldopam in dogs at differing doses and in dogs with clinical conditions such as acute nonpolyuric kidney injury is warranted.
Related Materials
Product #
Molecular Formula
Add to Cart
Fenoldopam mesylate, ≥98% (HPLC)
C16H16ClNO3 · CH4O3S
Fenoldopam monohydrobromide, ≥98%
C16H16ClNO3 · HBr