EMAIL THIS PAGE TO A FRIEND
The British journal of nutrition
Resveratrol attenuates steatosis in obese Zucker rats by decreasing fatty acid availability and reducing oxidative stress.
PMID 21733326
Abstract
Non-alcoholic fatty liver disease (NAFLD) is one of the most common manifestations of chronic liver disease worldwide. The aim of the present study was to assess the effect of resveratrol on liver fat accumulation, as well as on the activity of those enzymes involved in lipogenesis and fatty acid oxidation in fa/fa Zucker rats. A total of thirty rats were assigned to three experimental groups and orally treated with resveratrol for 6 weeks, or without resveratrol (C: control group; RSV15 group: 15 mg/kg body weight per d; RSV45 group: 45 mg/kg body weight per d). Liver histological analysis was performed by microscopy. Levels of hepatic carnitine palmitoyltransferase-Ia (CPT-Ia), acyl-coenzyme A oxidase (ACO), fatty acid synthase, glucose-6-phosphate dehydrogenase and malic enzyme were assessed by spectrophotometry, and acetyl-CoA carboxylase was assessed by radiometry. Commercial kits were used to determine serum TAG, NEFA, total HDL and non-HDL-cholesterol, glycerol, ketonic bodies, glucose, insulin, adiponectin, aspartate aminotransferase (AST), alanine aminotransferase (ALT) and alkaline phosphatase (ALP), hepatic TAG, thiobarbituric acid reactive substrates, GSH (GSSG) and superoxide dismutase. Resveratrol reduced liver weight and TAG content. It did not modify the activity of lipogenic enzymes but it did increase CPT-Ia and ACO activities. NEFA and ALP were reduced in both resveratrol-treated groups. AST/GOT was reduced only by the lowest dose. ALT/GPT, TAG and adiponectin remained unchanged. Resveratrol reduced liver oxidative stress. This study demonstrates that resveratrol can protect the liver from NAFLD by reducing fatty acid availability. Moreover, resveratrol also protects liver from oxidative stress.