EMAIL THIS PAGE TO A FRIEND
Fertility and sterility
Effect of combined metformin and oral contraceptive therapy on metabolic factors and endothelial function in overweight and obese women with polycystic ovary syndrome.
PMID 21733508
Abstract
In this randomized, double-blind, placebo-controlled study, 19 overweight women with polycystic ovary syndrome were randomized to a 3-month course of either metformin plus combined hormonal oral contraceptive (OC) (n = 9) or OC plus matched placebo (n = 10). After 3 months, both treatments had similar effects on androgen levels, lipid profile, insulin sensitivity, and serum inflammatory markers, but flow-mediated dilatation increased by 69.0% in the metformin plus OC group while it remained unchanged in the OC group. CLINICAL TRIAL REGISTRATION NO: NCT00682890.