EMAIL THIS PAGE TO A FRIEND
Planta
Isolation and characterization of the fragrant cyclamen O-methyltransferase involved in flower coloration.
PMID 21735197
Abstract
Anthocyanin O-methyltransferase (OMT) is one of the key enzymes for anthocyanin modification and flower pigmentation. We previously bred a novel red-purple-flowered fragrant cyclamen (KMrp) from the purple-flowered fragrant cyclamen 'Kaori-no-mai' (KM) by ion-beam irradiation. Since the major anthocyanins in KMrp and KM petals were delphinidin 3,5-diglucoside and malvidin 3,5-diglucoside, respectively, inactivation of a methylation step in the anthocyanin biosynthetic pathway was indicated in KMrp. We isolated and compared OMT genes expressed in KM and KMrp petals. RT-PCR analysis revealed that CkmOMT2 was expressed in the petals of KM but not in KMrp. Three additional CkmOMTs with identical sequences were expressed in petals of both KM and KMrp. Genomic PCR analysis revealed that CkmOMT2 was not amplified from the KMrp genome, indicating that ion-beam irradiation caused a loss of the entire CkmOMT2 region in KMrp. In vitro enzyme assay demonstrated that CkmOMT2 catalyzes the 3' or 3',5' O-methylation of the B-ring of anthocyanin substrates. These results suggest that CkmOMT2 is functional for anthocyanin methylation, and defective expression of CkmOMT2 is responsible for changes in anthocyanin composition and flower coloration in KMrp.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
43725
Delphinidin chloride, analytical standard
C15H11ClO7
68120
Malvidin chloride, ≥95.0% (HPLC)
C17H15ClO7