EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis and biological evaluation of 4β-acrylamidopodophyllotoxin congeners as DNA damaging agents.
PMID 21737288
Abstract
A series of new 4β-acrylamidopodophyllotoxin derivatives (13a-o) were synthesized by coupling of substituted acrylic acids (10a-l and 11m-o) to the 4β-aminopodophyllotoxin. The synthesized derivatives 13a-o were evaluated for their cytotoxicity against five human cancer cell lines (breast, oral, colon, lung and ovarian). These podophyllotoxin conjugates have shown promising activity with GI₅₀ values ranging from <0.1 to 0.29 μM. Some of the compounds 13j, 13k and 13l that showed significant antiproliferative activity were also evaluated for related cytotoxic effects in MCF-7 cells, and compared to etoposide. These compounds (13j, 13k and 13l) showed G2/M cell cycle arrest and the apoptotic event was found to be due to both the single-strand DNA breaks as observed by comet assay as well as double-strand breaks as observed by the large accumulation of gamma H2AX foci.