EMAIL THIS PAGE TO A FRIEND
Clinical cancer research : an official journal of the American Association for Cancer Research
Membranous expression of ectodomain isoforms of the epidermal growth factor receptor predicts outcome after chemoradiotherapy of lymph node-negative cervical cancer.
PMID 21737508
Abstract
We compared the prognostic significance of ectodomain isoforms of the epidermal growth factor receptor (EGFR), which lack the tyrosine kinase (TK) domain, with that of the full-length receptor and its autophosphorylation status in cervical cancers treated with conventional chemoradiotherapy. Expression of EGFR isoforms was assessed by immunohistochemistry in a prospectively collected cohort of 178 patients with squamous cell cervical carcinoma, and their detection was confirmed with Western blotting and reverse transcriptase PCR. A proximity ligation immunohistochemistry assay was used to assess EGFR-specific autophosphorylation. Pathways associated with the expression of ectodomain isoforms were studied by gene expression analysis with Illumina beadarrays in 110 patients and validated in an independent cohort of 41 patients. Membranous expression of ectodomain isoforms alone, without the coexpression of the full-length receptor, showed correlations to poor clinical outcome that were highly significant for lymph node-negative patients (locoregional control, P = 0.0002; progression-free survival, P < 0.0001; disease-specific survival, P = 0.005 in the log-rank test) and independent of clinical variables. The ectodomain isoforms were primarily 60-kD products of alternative EGFR transcripts. Their membranous expression correlated with transcriptional regulation of oncogenic pathways including activation of MYC and MAX, which was significantly associated with poor outcome. This aggressive phenotype of ectodomain EGFR expressing tumors was confirmed in the independent cohort. Neither total nor full-length EGFR protein level, or autophosphorylation status, showed prognostic significance. Membranous expression of ectodomain EGFR isoforms, and not TK activation, predicts poor outcome after chemoradiotherapy for patients with lymph node-negative cervical cancer.