EMAIL THIS PAGE TO A FRIEND
Anticancer research
Expression of iNOS--a favourable prognostic marker for early-stage carcinoma of the uterine cervix.
PMID 21737658
Abstract
In former studies, raised urine cGMP levels have been reported to predict adverse outcome in cervical cancer. The main objective of the present study was to investigate the value of nitric oxide synthase (iNOS) and other members of the cGMP pathway as potential biomarkers for prognosis of cervical carcinoma. Tissue samples from 85 patients surgically treated for early-stage cervical carcinoma were immunohistochemically stained for iNOS, soluble guanylyl cyclase subunits α1 (sGC-α1), soluble guanylyl cyclase α2 (sGC-α2), and phosphodiesterase 5, each sample evaluated microscopically by a semi-quantitative score. Results were correlated to recurrence and FIGO stage (depth of tumour cell infiltration). Correlation was found between high expression of iNOS in tumour cells and low risk of recurrence (p=0.019, p=0.05, p=0.022 and p=0.025). High expression of iNOS, sGC-α1 and sGC-α2 also correlated to superficial tumour growth. Our results demonstrate that iNOS expression in cervical tumour tissue is a robust prognostic marker for cervical cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
20357
ORBO 2-HMP on Supelpak 20N Tube, W,W,W separators, O.D. × L 8 mm × 115 mm, pkg of 25 ea
20257-U
ORBO 23 2-HMP on Amberlite® XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
20231
ORBO 24 2-HMP on Amberlite® XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
20832-U ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
20034 ORBO 403 Tenax® TA specially treated (60/80), 100/50 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 50 ea
20262 ORBO 42 Small Supelpak-20 specially treated Amberlite® XAD®-2 (20/40), 66/33 mg, W,F,F (specially treated) separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
20258 ORBO 43 Supelpak-20 specially treated Amberlite® XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
20260-U ORBO 44Supelpak-20 specially treated Amberlite® XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
20350
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite® XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
20030-U
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
20032
ORBO 506 Activated Silica Gel (45/60) 300/150 mg, W,W,W separators, O.D. × L 8 mm × 75 mm, pkg of 50 ea
20263
ORBO 52 Large Activated Silica Gel (45/60) 150/150 mg, W,W,W separators, O.D. × L 8 mm × 75 mm, pkg of 50 ea
20229
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
20265
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
20033
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
20049 ORBO 605 Amberlite® XAD®-2 specially cleaned (20/50) 100/50 mg, W,W,W separators, O.D. × L 6 mm × 100 mm, pkg of 50 ea
20050-U ORBO 608 Amberlite® XAD®-2 (20/50) 150/75 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
20051 ORBO 609 Amberlite® XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
20052 ORBO 613 Amberlite® XAD®-4 80/40 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
20029-U ORBO 65P (OVS) Amberlite® XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
20812-U ORBO 80 Coated Filter, with cassettes (unassembled)
20811
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
20069
ORBO 827 LpDNPH Coated Filter, 37mm, pkg of 25 ea
20362 ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate