EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Simple synthesis of 1,3-cyclopentanedione derived probes for labeling sulfenic acid proteins.
PMID 21738918
Abstract
Facile, two-step synthesis and kinetic characterization of new chemical probes for selective labeling of sulfenic acid (-SOH) in proteins are presented. The synthesis route relies on the simple and highly efficient Michael addition of thiol containing tags or linkers to 4-cyclopentene-1,3-dione, the unsaturated derivative of 1,3-cyclopentanedione.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
177164
1,3-Cyclopentanedione, 97%
C5H6O2
161683
4-Cyclopentene-1,3-dione, 95%
C5H4O2