EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Diversity-oriented production of metabolites derived from chorismate and their use in organic synthesis.
PMID 21739551
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1761
Chorismic acid from Enterobacter aerogenes, ≥80%
C10H10O6