EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Measurement of glycidol hemoglobin adducts in humans who ingest edible oil containing small amounts of glycidol fatty acid esters.
PMID 21740950
Abstract
Hemoglobin (Hb) adducts are frequently used to address and/or monitor exposure to reactive chemicals. Glycidol (G), a known animal carcinogen, has been reported to form Hb adducts. Here, we measure G adduct levels in humans who daily ingest DAG oil, an edible oil consisting mainly of diacylglycerol. Since DAG oil contains a small amount of glycidol fatty acid esters (GEs), possible exposure to G released from GEs has been raised as a possible concern. For measurement of Hb adducts, we employed the N-alkyl Edman method reported by Landin et al. (1996) using gas chromatography-tandem mass spectrometry with minor modifications to detect G-Hb adducts as N-(2,3-dihydroxy-propyl)valine (diHOPrVal). Blood samples were collected from 7 DAG oil users and 6 non-users, and then G-Hb adduct levels were measured. G-Hb adducts were detected in all samples. The average level of diHOPrVal was 3.5±1.9pmol/g globin in the DAG oil users and 7.1±3.1pmol/g globin in the non-users. We conclude that there is no increased exposure to G in individuals who daily ingest DAG oil.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
480819
(R)-(+)-Glycidol, 97%, optical purity ee: 98% (GLC)
C3H6O2
G5809
Glycidol, 96%
C3H6O2