EMAIL THIS PAGE TO A FRIEND
Journal of emergency nursing: JEN : official publication of the Emergency Department Nurses Association
Epinephrine in ventricular fibrillation: friend or foe? A review for the Emergency Nurse.
PMID 21741574