EMAIL THIS PAGE TO A FRIEND
Brain : a journal of neurology
Endogenous morphine-like compound immunoreactivity increases in parkinsonism.
PMID 21742735
Abstract
Morphine is endogenously synthesized in the central nervous system and endogenous dopamine is thought to be necessary for endogenous morphine formation. As Parkinson's disease results from the loss of dopamine and is associated with central pain, we considered how endogenous morphine is regulated in the untreated and l-DOPA-treated parkinsonian brain. However, as the cellular origin and overall distribution of endogenous morphine remains obscure in the pathological adult brain, we first characterized the distribution of endogenous morphine-like compound immunoreactive cells in the rat striatum. We then studied changes in the endogenous morphine-like compound immunoreactivity of medium spiny neurons in normal, Parkinson's disease-like and l-DOPA-treated Parkinson's disease-like conditions in experimental (rat and monkey) and human Parkinson's disease. Our results reveal an unexpected dramatic upregulation of neuronal endogenous morphine-like compound immunoreactivity and levels in experimental and human Parkinson's disease, only partially normalized by l-DOPA treatment. Our data suggest that endogenous morphine formation is more complex than originally proposed and that the parkinsonian brain experiences a dramatic upregulation of endogenous morphine immunoreactivity. The functional consequences of such endogenous morphine upregulation are as yet unknown, but based upon the current knowledge of morphine signalling, we hypothesize that it is involved in fatigue, depression and pain symptoms experienced by patients with Parkinson's disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
120693
α-Methyl-DL-tyrosine, 98%
C10H13NO3
M8131
α-Methyl-L-tyrosine, ≥98% (TLC)
C10H13NO3