EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Design and discovery of a selective small molecule κ opioid antagonist (2-methyl-N-((2'-(pyrrolidin-1-ylsulfonyl)biphenyl-4-yl)methyl)propan-1-amine, PF-4455242).
PMID 21744827
Abstract
By use of parallel chemistry coupled with physicochemical property design, a series of selective κ opioid antagonists have been discovered. The parallel chemistry strategy utilized key monomer building blocks to rapidly expand the desired SAR space. The potency and selectivity of the in vitro κ antagonism were confirmed in the tail-flick analgesia model. This model was used to build an exposure-response relationship between the κ K(i) and the free brain drug levels. This strategy identified 2-methyl-N-((2'-(pyrrolidin-1-ylsulfonyl)biphenyl-4-yl)methyl)propan-1-amine, PF-4455242, which entered phase 1 clinical testing and has demonstrated target engagement in healthy volunteers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PZ0206
PF-04455242 hydrochloride, ≥98% (HPLC)
C21H28N2O2S · HCl