EMAIL THIS PAGE TO A FRIEND
Journal of pineal research
Melatonin promotes adventitious root regeneration in in vitro shoot tip explants of the commercial sweet cherry rootstocks CAB-6P (Prunus cerasus L.), Gisela 6 (P. cerasus × P. canescens), and MxM 60 (P. avium × P. mahaleb).
PMID 21749439
Abstract
The objectives of this study were to test the effects of melatonin (N-acetyl-5-methoxytryptamine), a natural compound of edible plants on the rooting of certain commercial sweet cherry rootstocks. Shoot tip explants from previous in vitro cultures of the cherry rootstocks CAB-6P (Prunus cerasus L.), Gisela 6 (P. cerasus × P. canescens), and M × M 60 (P. avium × P. mahaleb) were included in the experiment. The effect of indole-3-acetic acid (IAA) and indole-3-butyric acid (IBA) alone or in combination with melatonin was tested concerning their rooting potential. Seven concentrations of melatonin (0, 0.05, 0.1, 0.5, 1, 5, and 10 μM) alone or in combination with 5.71 μM of IAA or 4.92 μM of IBA were tested. For each rootstock, 21 treatments were included. The explants were grown in glass tubes containing 10 mL of substrate. The parameters measured include rooting percentage, number of roots per rooted explant, root length, and callus formation. The data presented in this study show that melatonin has a rooting promoting effect at a low concentration but a growth inhibitory effect at high concentrations. In the absence of auxin, 1 μM melatonin had auxinic response concerning the number and length of roots, but 10 μM melatonin was inhibitory to rooting in all the tested rootstocks. The final conclusion of this experiment is that exogenously applied melatonin acted as a rooting promoter and its action was similar to that of IAA.