EMAIL THIS PAGE TO A FRIEND
ChemMedChem
Synthesis and structure-activity relationships of a class of sodium iodide symporter function inhibitors.
PMID 21751403
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
185493
N-Benzylaniline, ≥99%
C13H13N