EMAIL THIS PAGE TO A FRIEND
European journal of haematology
Efficacy and safety of the third-generation chloroethylnitrosourea fotemustine for the treatment of chemorefractory T-cell lymphomas.
PMID 21752099
Abstract
Patients with recurring T-cell non-Hodgkin lymphoma (T-NHL) are incurable and candidate for investigational agents. Here, we report on five patients with T-NHL refractory to multiple chemotherapy lines, including in all cases alkylators and gemcitabine, who received the third-generation chloroethylnitrosourea fotemustine at a dose of 120 mg/m(2) every 21 d, up to eight courses. Median actual dose intensity was 79%; toxicity was manageable and mainly hematological. One complete remission, one partial remission, two protracted disease stabilization, and one transient, minor response were achieved. Time to progression ranged from 48 to 240+ d. This is the first evidence ever reporting the activity of fotemustine in end-stage T-NHL. Formal studies with this agent are warranted in T-cell malignancies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F7307
Fotemustine, ≥98% (HPLC)
C9H19ClN3O5P