EMAIL THIS PAGE TO A FRIEND
The Journal of Nutritional Biochemistry
Postnatal early overnutrition dysregulates the intrarenal renin-angiotensin system and extracellular matrix-linked molecules in juvenile male rats.
PMID 21752621