EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Experimental and theoretical study on IR and NMR spectra of several tetrachlorinated diphenyl sulfides.
PMID 21752704
Abstract
Tetrachlorinated diphenyl sulfides (TCDPSs) are environmentally interesting compounds. In this paper, both experimental and theoretical studies on IR and (1)H NMR as well as (13)C NMR chemical shifts of 4 synthesized TCDPSs have been carried out. The optimized geometries were obtained at the B3LYP/6-311G** level by using the Gaussian 03 program. Meanwhile, related spectral parameters were calculated. In addition, the experimental values were compared with the calculated ones. The results show that the scaled theoretical vibration frequencies are in good accordance with the observations, and computed chemical shifts are consistent with the experimental values.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P35316
Diphenyl sulfide, 98%
C12H10S
46528
Diphenyl sulfide, PESTANAL®, analytical standard
C12H10S