EMAIL THIS PAGE TO A FRIEND
Medical principles and practice : international journal of the Kuwait University, Health Science Centre
Postoperative analgesia in impacted third molar surgery: the role of preoperative diclofenac sodium, paracetamol and lornoxicam.
PMID 21757939
Abstract
The aim of this study was to compare the postoperative analgesic effects of preoperative intravenous (i.v.) paracetamol, diclofenac sodium and lornoxicam (nonsteroidal anti-inflammatory drugs). Sixty patients with impacted third molar who underwent surgical removal were randomly allocated into three groups: group P (n = 20), group D (n = 20) and group L (n = 20). Group P received preoperatively 1 g paracetamol i.v., group D 75 mg diclofenac sodium i.m. and group L 8 mg lornoxicam i.v. Postoperative pain intensity, additional consumption of analgesics postoperatively and postoperative complications were compared among groups. The groups were comparable for pain scores (p > 0.05). Maximum pain scores were recorded in postoperative 4th h in all groups (group L 22, 14-44 mm; group P 24, 13-43 mm; group D 14, 10-24 mm, p = 0.117). Patients experienced high satisfaction scores which were comparable among groups (group L 85, 75-100 mm; group P 87, 70-95 mm; group D 84, 77-98 mm, p = 0.457). Preoperative intramuscular diclofenac, intravenous paracetamol and lornoxicam effectively decreased the pain scores. The patients were satisfied with the three postoperative pain management regimens.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0338
Lornoxicam, ≥98% (HPLC)
C13H10ClN3O4S2