EMAIL THIS PAGE TO A FRIEND
Pharmacology
Pharmacokinetics of fluoxetine in rhesus macaques following multiple routes of administration.
PMID 21757974
Abstract
Fluoxetine (Prozac) is a selective serotonin reuptake inhibitor currently used to treat depression and mood disorders. It has been widely studied clinically and preclinically, yet there is limited knowledge of its pharmacokinetics in nonhuman primates. The present study characterized the pharmacokinetics of fluoxetine and its active metabolite norfluoxetine in rhesus macaques following both acute (1, 3, 5.6 and 10 mg/kg) and chronic doses (5.6 and 10 mg/kg/day) via different routes of administration (intravenous, subcutaneous, intramuscular, and oral). Blood samples were collected at multiple time points following administration and analyzed using mass spectrometry. Fluoxetine had a half-life of 11-16 h and norfluoxetine had a half-life of 21-29 h. Potentially functionally significant serum concentrations of norfluoxetine were present at 24 h even after a single administration of fluoxetine. Similar to observations in humans under steady state conditions, norfluoxetine accounted for the greater percentage of active drug in the blood stream. A daily dose of 10 mg/kg administered orally maintained serum concentrations in the human clinical range over the course of 6 weeks. Given the long half-lives of fluoxetine and norfluoxetine observed in this study, precautions should be taken when designing preclinical studies to prevent accumulation of drug serum concentrations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F133
Norfluoxetine hydrochloride, ≥97%, solid
C16H16F3NO · HCl