EMAIL THIS PAGE TO A FRIEND
Journal of Neuropathology and Experimental Neurology
Peripheral neuropathy is linked to a severe form of myotonic dystrophy in transgenic mice.
PMID 21760538
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB144P
Anti-Choline Acetyltransferase Antibody, clone, Chemicon®, from goat