EMAIL THIS PAGE TO A FRIEND
Developmental dynamics : an official publication of the American Association of Anatomists
Analysis of Cdh22 expression and function in the developing mouse brain.
PMID 21761482
Abstract
Classical cadherins are important cell adhesion molecules specifying and separating brain nuclei and developmental compartments. Cadherin-22 (Cdh22) belongs to type II subfamily of classical cadherins, and is expressed at the midbrain-hindbrain boundary during early embryogenesis. In Fgfr1 mutant mouse embryos, which have a disturbed midbrain-hindbrain border, Cdh22 is down-regulated. Here, we studied expression of Cdh22 in developing mouse brain in more detail and compared it to expression of related family members. This revealed both complementary and overlapping patterns of Cdh22, Cdh11, Cdh8, and Cdh6 expression in distinct regions of the forebrain and midbrain. We used a mutated allele of Cdh22 to study its function in brain development. Loss of Cdh22 caused reduced postnatal viability. Despite strong Cdh22 expression in the developing brain, we did not observe defects in compartmentalization or abnormalities in the midbrain and forebrain nuclei in Cdh22 mutants. This may be explained by functional redundancy between type II cadherins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV49648
Anti-CDH22 antibody produced in rabbit, IgG fraction of antiserum