EMAIL THIS PAGE TO A FRIEND
Rinsho byori. The Japanese journal of clinical pathology
[Synthetic protease inhibitors for the treatment of DIC].
PMID 21761764
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G2417
Gabexate mesylate, analytical standard, for drug analysis
C16H23N3O4 · CH4O3S