EMAIL THIS PAGE TO A FRIEND
Biomacromolecules
Cationic amphiphilic star and linear block copolymers: synthesis, self-assembly, and in vitro gene transfection.
PMID 21761847
Abstract
A series of amphiphilic star and linear block copolymers were synthesized using ATRP. The core consisted of either polystyrene (PS) or poly(n-butyl acrylate) (PBuA), having different glass-transition (T(g)) values. These polymers were used as macroinitiators in the polymerization of the cationic 2-(dimethylamino)ethyl methacrylate (DMAEMA). The polymers were used to study the effects of polymer architecture and flexibility on the self-assembling properties, DNA complexation, and transfection. All polymers formed core-shell micelles in aqueous solutions and condensed plasmid DNA. Linear PDMAEMA-PBuA-PDMAEMA has transfection efficiency comparable to PEI25K in ARPE19 cell line. Glassy state of the micellar core and star-shaped architecture decreased the DNA transfection compared with the rubbery and linear polymer structures. The polymers showed low cellular toxicity at low nitrogen/phosphate (n/p) ratios.
Related Materials