EMAIL THIS PAGE TO A FRIEND
Journal of the American Academy of Dermatology
Primary cutaneous trichosporonosis caused by Trichosporon dermatis in an immunocompetent man.
PMID 21763573
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D16606
6,9-Diamino-2-ethoxyacridine-DL-lactate monohydrate, 99%
C15H15N3O · C3H6O3 · H2O