EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Design and synthesis of 2-phenylimidazo[1,2-a]pyridines as a novel class of melatonin receptor ligands.
PMID 21764185
Abstract
A novel class of imidazo[1,2-a]pyridines as melatonin receptor ligands is designed and synthesized. The affinities of 3-(6-methoxy-2-phenylimidazo[1,2-a]pyridine-3-yl)-N-methyl-propionamide 8, N-[2-(6-methoxy-2-phenylimidazo[1,2-a]pyridine-3-yl)-ethyl]-acetamide 13 and N-(1-hydroxy-3-(5-methoxy-2-phenyl-1H-indol-3-yl)propan-2-yl)acetamide 18 are evaluated for binding on melatonin receptors. Compound 8 present good selectivity for MT(2) over MT(1) (MT(1)/MT(2) = 19) and compound 13 have good affinities for both MT(1) (Ki :28 nM) and MT(2) (Ki : 8 nM).