EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Opioid δ₁ receptor antagonist 7-benzylidenenaltrexone as an effective resistance reverser for chloroquine-resistant Plasmodium chabaudi.
PMID 21764311
Abstract
We evaluated antimalarial and/or chloroquine-resistance reversing effects of five opioid receptor antagonists. Although none of the evaluated compounds showed antimalarial effects, some of them, especially the δ(1) receptor antagonist, 7-benzylidenenaltrexone (BNTX) exhibited potent chloroquine-resistance reversing effects in Plasmodium chabaudi.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B8312
BNTX maleate salt hydrate, ≥98% (HPLC)
C31H31NO8 · xH2O