EMAIL THIS PAGE TO A FRIEND
Canadian journal of gastroenterology = Journal canadien de gastroenterologie
An evaluation of the use of serum 7-alpha-hydroxycholestenone as a diagnostic test of bile acid malabsorption causing watery diarrhea.
PMID 21766092
Abstract
Bile acid malabsorption (BAM) is a recognized cause of watery diarrhea, often diagnosed empirically based on clinical response to cholestyramine. The radionuclide selenium-labelled homocholic acid-taurine whole body retention test is expensive, labour intensive and of limited availability. To report on the clinical performance of serum 7-alphahydroxy-4-cholesten-3-one (7HCO) as a test of BAM in adult patients with unexplained diarrhea. Patients with unexplained diarrhea were investigated over a three-year period. Final diagnosis was determined based on medical history and investigations, serum levels of 7HCO and response to cholestyramine. ROC analysis was used to determine the ideal upper reference range cut-off value to optimize sensitivity/specificity for BAM. Time of blood specimen collection was recorded to investigate possible variation in results throughout the working day. ROC analysis yielded a sensitivity/specificity of 90%/77% for type 1 BAM (ileal disease/resection) and 97%/74% for type 2 BAM (idiopathic) using 30 ng/mL as the upper limit of normal for serum 7HCO when compared with all other patients. Of 813 patients, 196 tested positive. Serum 7HCO levels were significantly higher in blood specimens that were collected between 12:00 and 13:00 (median 24 ng/mL) than in specimens collected between 09:00 and 10:00 (median 17 ng/mL) (P<0.05). Serum 7HCO testing is a simple, sensitive, noninvasive, inexpensive alternative to other more commonly used tests for BAM. Time of specimen collection, however, resulted in small but significant result variations and, although unlikely to have much impact on test value, it should ideally be standardized.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
94145
7α-Hydroxy-4-cholesten-3-one-25,26,26,26,27,27,27-d7, ≥95.0% (HPLC)
C27D7H37O2