EMAIL THIS PAGE TO A FRIEND
Nihon rinsho. Japanese journal of clinical medicine
[Sulfonylureas and glinides].
PMID 21766676
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N3538
Nateglinide, ≥98% (HPLC), solid
C19H27NO3