EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Ditopic triply hydrogen-bonded heterodimers.
PMID 21769372
Abstract
The synthesis and self-assembly of a stable hydrogen-bonded heterodimer comprising ditopic ureidoimidazole and amidoisocytosine motifs is described. The heterodimer appears to exhibit high stability in deuterochloroform as evidenced by (1)H NMR, DOSY and (1)H-(1)H ROESY.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2127
Isocytosine, ≥99%
C4H5N3O